siRNA Details
Gene Symbol: TAPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003190.4 | NP_003181.3 | 2 | 428 | ||
NM_172208.2 | NP_757345.2 | 2 | 428 | |||
NM_172209.2 | NP_757346.2 | 2 | 428 | |||
XM_011514828.2 | XP_011513130.1 | 2 | 295 | |||
XM_017011227.1 | XP_016866716.1 | 2 | 295 | |||
GenBank mRNA | AB010639.1 | - | 2 | 95 | ||
AF009510.1 | - | 2 | 98 | |||
AF029750.1 | - | 2 | 120 | |||
AF067286.3 | - | 2 | 90 | |||
AF314222.1 | - | 2 | 82 | |||
AU117273.1 | - | 2 | 295 | |||
BC064966.1 | - | 2 | 285 | |||
BC080574.1 | - | 2 | 253 | 6572933 | ||
BI523615.1 | - | 2 | 428 | |||
EU693375.1 | - | 2 | 82 | |||
Y13582.1 | - | 2 | 211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
