siRNA Details
Gene Symbol: HIST1H3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021018.2 | NP_066298.1 | 1 | 17 | ||
GenBank mRNA | AK314142.1 | - | 1 | 50 | ||
BC069818.1 | - | 1 | 17 | 7262721 | ||
CR541983.1 | - | 1 | 15 | |||
CR542011.1 | - | 1 | 15 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
