siRNA Details
Gene Symbol: ERBIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006600.2 | NP_001006600.1 | 22 | 4016 | ||
NM_001253697.1 | NP_001240626.1 | 24 | 4346 | |||
NM_001253698.1 | NP_001240627.1 | 23 | 4223 | |||
NM_001253699.1 | NP_001240628.1 | 24 | 4367 | |||
NM_001253701.1 | NP_001240630.1 | 23 | 4211 | |||
NM_018695.3 | NP_061165.1 | 23 | 4223 | |||
XM_005248554.2 | XP_005248611.1 | 25 | 4445 | |||
XM_005248555.3 | XP_005248612.1 | 25 | 4433 | |||
XM_017009635.1 | XP_016865124.1 | 25 | 4261 | |||
XM_017009636.1 | XP_016865125.1 | 24 | 4310 | |||
XM_017009637.1 | XP_016865126.1 | 22 | 3959 | |||
GenBank mRNA | AB033051.2 | - | 21 | 3913 | ||
AF263744.1 | - | 23 | 4236 | |||
AF276423.1 | - | 24 | 4179 | |||
AK027077.1 | - | 2 | 288 | |||
AK295708.1 | - | 17 | 1927 | |||
AK304693.1 | - | 23 | 4169 | |||
BC115012.1 | - | 22 | 3993 | 7961934 | ||
BC115015.1 | - | 22 | 3981 | 7961940 | ||
BC126464.1 | - | 23 | 4004 | 8992180 | ||
BC144075.1 | - | 24 | 4148 | 9052595 | ||
BU145959.1 | - | 2 | 637 | |||
CB998970.1 | - | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
