siRNA Details
Gene Symbol: ERBIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006600.2 | NP_001006600.1 | 24 | 4934 | ||
NM_001253697.1 | NP_001240626.1 | 26 | 5264 | |||
NM_001253698.1 | NP_001240627.1 | 24 | 5066 | |||
NM_001253699.1 | NP_001240628.1 | 26 | 5285 | |||
NM_001253701.1 | NP_001240630.1 | 25 | 5129 | |||
NM_018695.3 | NP_061165.1 | 25 | 5141 | |||
XM_005248554.2 | XP_005248611.1 | 27 | 5363 | |||
XM_005248555.3 | XP_005248612.1 | 27 | 5351 | |||
XM_017009635.1 | XP_016865124.1 | 27 | 5179 | |||
XM_017009636.1 | XP_016865125.1 | 26 | 5228 | |||
XM_017009637.1 | XP_016865126.1 | 24 | 4877 | |||
GenBank mRNA | AB033051.2 | - | 23 | 4831 | ||
AF263744.1 | - | 26 | 5117 | |||
AK027077.1 | - | 4 | 1206 | |||
AK074288.1 | - | 2 | 858 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
