siRNA Details
Gene Symbol: SMAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001419.2 | NP_001001419.1 | 4 | 768 | ||
NM_001001420.2 | NP_001001420.1 | 2 | 609 | |||
NM_005903.6 | NP_005894.3 | 3 | 684 | |||
XM_017009470.1 | XP_016864959.1 | 4 | 900 | |||
GenBank mRNA | AF009678.1 | - | 1 | 336 | ||
AF010601.1 | - | 1 | 453 | |||
AK313397.1 | - | 3 | 684 | |||
BC009682.2 | - | 1 | 490 | 3906006 | ||
BX510058.1 | - | 2 | 610 | |||
BX648328.1 | - | 3 | 623 | |||
CR457424.1 | - | 1 | 322 | |||
CR749366.1 | - | 3 | 623 | |||
CR749473.1 | - | 3 | 611 | |||
U59913.1 | - | 1 | 452 | |||
U73825.1 | - | 1 | 471 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
