siRNA Details
Gene Symbol: SMAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001419.2 | NP_001001419.1 | 9 | 3923 | ||
NM_001001420.2 | NP_001001420.1 | 7 | 3764 | |||
NM_005903.6 | NP_005894.3 | 8 | 3839 | |||
XM_017009470.1 | XP_016864959.1 | 9 | 4055 | |||
GenBank mRNA | AK055211.1 | - | 1 | 385 | ||
BX648328.1 | - | 8 | 3779 | |||
CR749473.1 | - | 8 | 3764 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
