siRNA Details
Gene Symbol: SLC7A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020949.2 | NP_066000.2 | 6 | 1266 | ||
GenBank mRNA | AB046833.1 | - | 5 | 717 | ||
AK122655.1 | - | 6 | 1261 | |||
BC022968.1 | - | 5 | 882 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
