siRNA Details
Gene Symbol: ABTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032548.3 | NP_115937.1 | 7 | 644 | ||
NM_172027.2 | NP_742024.1 | 7 | 633 | |||
NR_033429.1 | 7 | 598 | ||||
XM_006713769.3 | XP_006713832.1 | 7 | 667 | |||
XM_011513213.1 | XP_011511515.1 | 7 | 567 | |||
XM_017007287.1 | XP_016862776.1 | 7 | 604 | |||
GenBank mRNA | AB053324.1 | - | 7 | 623 | ||
AB053325.1 | - | 7 | 612 | |||
AB053326.1 | - | 7 | 577 | |||
AF297986.1 | - | 7 | 590 | |||
AF447886.1 | - | 6 | 734 | |||
AK090422.1 | - | 2 | 1396 | |||
AK315148.1 | - | 7 | 613 | |||
BC011858.2 | - | 7 | 631 | 4309664 | ||
BF664138.1 | - | 7 | 631 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
