siRNA Details
Gene Symbol: PARD3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302769.1 | NP_001289698.1 | 22 | 3452 | ||
NM_057177.6 | NP_476518.4 | 21 | 3245 | |||
NM_152526.5 | NP_689739.4 | 21 | 3266 | |||
NM_205863.3 | NP_995585.2 | 21 | 3149 | |||
XM_011510552.2 | XP_011508854.1 | 25 | 3795 | |||
XM_017003283.1 | XP_016858772.1 | 21 | 3742 | |||
XM_017003284.1 | XP_016858773.1 | 22 | 6788 | |||
XM_017003285.1 | XP_016858774.1 | 23 | 8339 | |||
XM_017003286.1 | XP_016858775.1 | 21 | 6684 | |||
XM_017003287.1 | XP_016858776.1 | 24 | 8521 | |||
XM_017003288.1 | XP_016858777.1 | 23 | 8220 | |||
XM_017003289.1 | XP_016858778.1 | 19 | 2899 | |||
XM_017003290.1 | XP_016858779.1 | 23 | 3399 | |||
XM_017003291.1 | XP_016858780.1 | 22 | 3271 | |||
XM_017003292.1 | XP_016858781.1 | 22 | 3333 | |||
XM_017003293.1 | XP_016858782.1 | 19 | 2837 | |||
GenBank mRNA | AB073472.1 | - | 23 | 2942 | ||
AB092439.1 | - | 24 | 3245 | |||
AF428250.1 | - | 22 | 3245 | |||
AF428251.1 | - | 21 | 3059 | |||
AF466152.1 | - | 21 | 3038 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
