siRNA Details
Gene Symbol: HES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019089.4 | NP_061962.2 | 1 | 134 | ||
GenBank mRNA | AK091122.1 | - | 1 | 134 | ||
BC012091.1 | - | 2 | 251 | |||
BC132698.1 | - | 1 | 97 | 40146720 | ||
BC136963.1 | - | 1 | 97 | 9020955 | ||
BC142687.1 | - | 1 | 114 | 40148619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
