siRNA Details
Gene Symbol: COLEC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255982.1 | NP_001242911.1 | 5 | 671 | ||
NM_001255983.1 | NP_001242912.1 | 5 | 671 | |||
NM_001255984.1 | NP_001242913.1 | 4 | 599 | |||
NM_001255985.1 | NP_001242914.1 | 7 | 633 | |||
NM_001255986.1 | NP_001242915.1 | 5 | 442 | |||
NM_001255987.1 | NP_001242916.1 | 4 | 370 | |||
NM_001255988.1 | NP_001242917.1 | 4 | 370 | |||
NM_001255989.1 | NP_001242918.1 | 3 | 298 | |||
NM_024027.4 | NP_076932.1 | 6 | 743 | |||
NM_199235.2 | NP_954705.1 | 7 | 822 | |||
NR_045659.1 | 7 | 936 | ||||
XM_005263853.4 | XP_005263910.1 | 5 | 365 | |||
XM_006711897.3 | XP_006711960.1 | 5 | 365 | |||
GenBank mRNA | AB119525.1 | - | 6 | 629 | ||
AB119650.1 | - | 5 | 557 | |||
AB119651.1 | - | 5 | 557 | |||
AB119652.1 | - | 4 | 485 | |||
AB119684.1 | - | 5 | 442 | |||
AB119685.1 | - | 4 | 370 | |||
AB119686.1 | - | 4 | 370 | |||
AB119687.1 | - | 3 | 298 | |||
AK303824.1 | - | 7 | 633 | |||
AK313840.1 | - | 6 | 745 | |||
AY358439.1 | - | 6 | 446 | |||
BC000078.2 | - | 6 | 453 | 3507377 | ||
BC009951.2 | - | 7 | 605 | 4125795 | ||
BM547424.1 | - | 6 | 614 | |||
CR936641.1 | - | 4 | 234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
