siRNA Details
Gene Symbol: SLC8A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015063.2 | NP_055878.1 | 10 | 3897 | ||
XM_005259172.1 | XP_005259229.1 | 9 | 3688 | |||
XM_017027159.1 | XP_016882648.1 | 7 | 2045 | |||
GenBank mRNA | AB029010.1 | - | 10 | 3712 | ||
AL512764.1 | - | 9 | 3086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
