siRNA Details
Gene Symbol: DDX43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018665.2 | NP_061135.2 | 13 | 1889 | ||
XM_011535926.1 | XP_011534228.1 | 12 | 1757 | |||
XM_011535927.2 | XP_011534229.1 | 12 | 1346 | |||
XM_011535928.2 | XP_011534230.1 | 12 | 1535 | |||
XM_017011003.1 | XP_016866492.1 | 12 | 1475 | |||
GenBank mRNA | AJ278110.1 | - | 13 | 1752 | ||
AK313686.1 | - | 13 | 1589 | |||
AL136751.1 | - | 13 | 1577 | |||
BC066938.1 | - | 13 | 1570 | 4829072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
