siRNA Details
Gene Symbol: DDX43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018665.2 | NP_061135.2 | 1, 2 | 599 | ||
XM_011535926.1 | XP_011534228.1 | 1, 2 | 599 | |||
XM_017011003.1 | XP_016866492.1 | 1, 2 | 267 | |||
GenBank mRNA | AJ278110.1 | - | 1, 2 | 462 | ||
AK303324.1 | - | 1, 2 | 308 | |||
AK313686.1 | - | 1, 2 | 299 | |||
AL136751.1 | - | 1, 2 | 287 | |||
BC066938.1 | - | 1, 2 | 280 | 4829072 | ||
BX646425.1 | - | 1, 2 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
