siRNA Details
Gene Symbol: DDX43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018665.2 | NP_061135.2 | 7 | 1200 | ||
XM_011535926.1 | XP_011534228.1 | 6 | 1068 | |||
XM_011535927.2 | XP_011534229.1 | 6 | 657 | |||
XM_011535928.2 | XP_011534230.1 | 6 | 846 | |||
XM_017011003.1 | XP_016866492.1 | 6 | 786 | |||
GenBank mRNA | AJ278110.1 | - | 7 | 1063 | ||
AK303324.1 | - | 6 | 827 | |||
AK313686.1 | - | 7 | 900 | |||
AL136751.1 | - | 7 | 888 | |||
BC066938.1 | - | 7 | 881 | 4829072 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
