siRNA Details
Gene Symbol: PCDHB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018930.3 | NP_061753.1 | 1 | 3088 | ||
GenBank mRNA | AF131761.1 | - | 1 | 1911 | ||
AF217748.1 | - | 1 | 2914 | |||
AK056391.1 | - | 1 | 3098 | |||
AY358720.1 | - | 1 | 3092 | |||
BC031837.1 | - | 1 | 3088 | 4931995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
