siRNA Details
Gene Symbol: LYZL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032517.4 | NP_115906.3 | 5 | 610 | ||
GenBank mRNA | AY358694.1 | - | 5 | 555 | ||
BC021730.2 | - | 5 | 576 | 4825273 | ||
BG209290.1 | - | 2 | 121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
