siRNA Details
Gene Symbol: PRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005039.3 | NP_005030.2 | 4 | 1109 | ||
NM_199353.2 | NP_955385.1 | 5 | 710 | |||
NM_199354.2 | NP_955386.1 | 5 | 650 | |||
GenBank mRNA | K03204.1 | - | 4 | 1109 | ||
K03205.1 | - | 5 | 698 | |||
K03206.1 | - | 5 | 646 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
