siRNA Details
Gene Symbol: PRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005039.3 | NP_005030.2 | 4 | 1085 | ||
NM_199353.2 | NP_955385.1 | 5 | 686 | |||
NM_199354.2 | NP_955386.1 | 5 | 626 | |||
GenBank mRNA | BC044827.1 | - | 4 | 1071 | ||
K03204.1 | - | 4 | 1085 | |||
K03205.1 | - | 5 | 674 | |||
K03206.1 | - | 5 | 622 |
Gene Symbol: PRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006248.3 | NP_006239.3 | 4 | 1338 | ||
GenBank mRNA | K03208.1 | - | 2 | 808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
