siRNA Details
Gene Symbol: TFG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007565.2 | NP_001007566.1 | 2 | 215 | ||
NM_001195478.1 | NP_001182407.1 | 2 | 120 | |||
NM_001195479.1 | NP_001182408.1 | 2 | 464 | |||
NM_006070.5 | NP_006061.2 | 2 | 464 | |||
XM_005247066.1 | XP_005247123.1 | 2 | 221 | |||
XM_006713472.1 | XP_006713535.1 | 2 | 185 | |||
XM_006713473.1 | XP_006713536.1 | 2 | 171 | |||
XM_011512334.1 | XP_011510636.1 | 2 | 399 | |||
XM_017005527.1 | XP_016861016.1 | 2 | 185 | |||
XM_017005528.1 | XP_016861017.1 | 2 | 171 | |||
XM_017005529.1 | XP_016861018.1 | 2 | 211 | |||
XM_017005530.1 | XP_016861019.1 | 2 | 399 | |||
GenBank mRNA | AB731569.1 | - | 1 | 30 | ||
AB731570.1 | - | 1 | 30 | |||
AK093456.1 | - | 2 | 370 | |||
BC001483.2 | - | 2 | 120 | 3507440 | ||
BC009241.2 | - | 2 | 343 | 2962943 | ||
BC023599.2 | - | 2 | 102 | 4555507 | ||
BC041600.1 | - | 2 | 189 | 5458899 | ||
BE409447.1 | - | 2 | 188 | |||
BT007428.1 | - | 1 | 30 | |||
CR456781.1 | - | 1 | 30 | |||
DA704111.1 | - | 2 | 221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
