siRNA Details
Gene Symbol: ITPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002223.3 | NP_002214.2 | 3 | 684 | ||
XM_017019266.1 | XP_016874755.1 | 3 | 683 | |||
XM_017019267.1 | XP_016874756.1 | 3 | 207 | |||
XM_017019268.1 | XP_016874757.1 | 1 | 27 | |||
XM_017019269.1 | XP_016874758.1 | 3 | 683 | |||
GenBank mRNA | AB012610.1 | - | 3 | 267 | ||
AK307712.1 | - | 3 | 560 | |||
AK310504.1 | - | 3 | 424 | |||
AY429550.1 | - | 2 | 201 | |||
D26350.1 | - | 3 | 685 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
