siRNA Details
Gene Symbol: H2BFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164416.1 | NP_001157888.1 | 1 | 131 | ||
XM_006724640.2 | XP_006724703.1 | 1 | 164 | |||
XM_011530922.2 | XP_011529224.1 | 1 | 327 | |||
GenBank mRNA | AK093522.1 | - | 1 | 131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
