siRNA Details
Gene Symbol: GTSF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008901.1 | NP_001008901.1 | 1 | 322 | ||
NM_176791.3 | NP_789761.1 | 1 | 322 | |||
XM_005260298.4 | XP_005260355.1 | 1 | 455 | |||
GenBank mRNA | AK058060.1 | - | 1 | 318 | ||
BC040049.1 | - | 1 | 247 | 5744556 | ||
BQ428678.1 | - | 1 | 326 | |||
BX355734.2 | - | 1 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
