siRNA Details
Gene Symbol: SEMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003007.4 | NP_002998.1 | 2 | 1314 | ||
GenBank mRNA | BC007096.1 | - | 3 | 1065 | 4251523 | |
BC055416.1 | - | 3 | 1065 | 6668871 | ||
BT007177.1 | - | 3 | 1044 | |||
J04440.1 | - | 2 | 1230 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
