siRNA Details
Gene Symbol: PSG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206650.1 | NP_001193579.1 | 5 | 1602 | ||
NM_001290042.1 | NP_001276971.1 | 6 | 1968 | |||
NM_002783.2 | NP_002774.2 | 6 | 1968 | |||
GenBank mRNA | BC030979.1 | - | 6 | 1963 | ||
U18467.1 | - | 6 | 1918 |
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 5 | 1819 | ||
NM_001316339.1 | NP_001303268.1 | 5 | 1732 | |||
NM_002780.4 | NP_002771.2 | 6 | 2098 | |||
NM_213633.2 | NP_998798.1 | 5 | 1819 | |||
XM_017026996.1 | XP_016882485.1 | 7 | 2160 | |||
XM_017027000.1 | XP_016882489.1 | 6 | 1885 | |||
GenBank mRNA | AF289606.1 | - | 6 | 1439 | ||
AK075149.1 | - | 6 | 1713 | |||
BC008405.1 | - | 8 | 1957 | 4043484 | ||
BC063127.1 | - | 6 | 1966 | 4768860 | ||
M33665.1 | - | 4 | 1239 | |||
M94891.1 | - | 6 | 1946 | |||
U18468.1 | - | 6 | 1865 | |||
U18469.1 | - | 5 | 1608 | |||
X17097.1 | - | 6 | 1953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
