siRNA Details
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 2 | 452 | ||
NM_002780.4 | NP_002771.2 | 2 | 452 | |||
NM_213633.2 | NP_998798.1 | 2 | 452 | |||
XM_017026995.1 | XP_016882484.1 | 3 | 515 | |||
XM_017026996.1 | XP_016882485.1 | 3 | 514 | |||
XM_017026997.1 | XP_016882486.1 | 2 | 346 | |||
XM_017026998.1 | XP_016882487.1 | 3 | 519 | |||
XM_017026999.1 | XP_016882488.1 | 3 | 520 | |||
XM_017027000.1 | XP_016882489.1 | 3 | 518 | |||
XM_017027001.1 | XP_016882490.1 | 2 | 346 | |||
XM_017027002.1 | XP_016882491.1 | 2 | 346 | |||
GenBank mRNA | AK075149.1 | - | 3 | 346 | ||
BC063127.1 | - | 2 | 320 | 4768860 | ||
BX403624.2 | - | 2 | 304 | |||
DA837940.1 | - | 2 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
