siRNA Details
Gene Symbol: HIF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022462.4 | NP_071907.4 | 4 | 478 | ||
NM_152794.3 | NP_690007.1 | 5 | 574 | |||
NM_152795.3 | NP_690008.2 | 5 | 542 | |||
NM_152796.2 | NP_690009.1 | 4 | 478 | |||
XM_005259152.4 | XP_005259209.1 | 6 | 670 | |||
XM_005259153.3 | XP_005259210.2 | 3 | 341 | |||
XM_005259155.4 | XP_005259212.1 | 6 | 670 | |||
XM_005259156.4 | XP_005259213.1 | 6 | 670 | |||
XM_017027132.1 | XP_016882621.1 | 6 | 670 | |||
XM_017027133.1 | XP_016882622.1 | 6 | 670 | |||
XM_017027134.1 | XP_016882623.1 | 5 | 540 | |||
XM_017027135.1 | XP_016882624.1 | 5 | 574 | |||
XM_017027136.1 | XP_016882625.1 | 6 | 670 | |||
XM_017027137.1 | XP_016882626.1 | 5 | 574 | |||
XM_017027138.1 | XP_016882627.1 | 5 | 540 | |||
XM_017027139.1 | XP_016882628.1 | 4 | 464 | |||
XM_017027140.1 | XP_016882629.1 | 4 | 394 | |||
XM_017027141.1 | XP_016882630.1 | 4 | 436 | |||
XM_017027142.1 | XP_016882631.1 | 4 | 464 | |||
GenBank mRNA | AF463492.1 | - | 5 | 537 | ||
AK299699.1 | - | 3 | 388 | |||
BC080551.1 | - | 5 | 513 | 6250259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
