siRNA Details
Gene Symbol: MT1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301267.1 | NP_001288196.1 | 3 | 316 | ||
NM_005950.2 | NP_005941.1 | 3 | 313 | |||
GenBank mRNA | BC020757.1 | - | 3 | 316 | 4767886 | |
BC048334.1 | - | 1 | 1229 | |||
BM817643.1 | - | 3 | 316 | |||
CD175975.1 | - | 3 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
