siRNA Details
Gene Symbol: ATP5S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003803.2 | NP_001003803.1 | 3 | 980 | ||
NM_001003805.2 | NP_001003805.1 | 3 | 980 | |||
NM_015684.3 | NP_056499.2 | 3 | 980 | |||
NR_033761.1 | 4 | 1016 | ||||
XM_005267537.3 | XP_005267594.1 | 3 | 887 | |||
XM_011536649.2 | XP_011534951.1 | 3 | 869 | |||
XM_011536650.2 | XP_011534952.1 | 3 | 871 | |||
XM_011536651.2 | XP_011534953.1 | 3 | 871 | |||
XM_011536652.1 | XP_011534954.1 | 3 | 269 | |||
XM_011536653.2 | XP_011534955.1 | 4 | 311 | |||
XM_011536654.2 | XP_011534956.1 | 3 | 875 | |||
XM_011536655.2 | XP_011534957.1 | 3 | 875 | |||
XM_011536656.2 | XP_011534958.1 | 3 | 907 | |||
XM_011536657.2 | XP_011534959.1 | 3 | 418 | |||
XM_017021220.1 | XP_016876709.1 | 3 | 351 | |||
GenBank mRNA | AF052186.1 | - | 3 | 932 | ||
AF390029.1 | - | 2 | 204 | |||
AK290008.1 | - | 3 | 668 | |||
AY052377.1 | - | 3 | 403 | |||
BC011549.1 | - | 3 | 980 | 4554461 | ||
BC087851.1 | - | 4 | 785 | |||
BG699167.1 | - | 3 | 316 | |||
BU189283.1 | - | 3 | 291 | |||
U79253.1 | - | 3 | 931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
