siRNA Details
Gene Symbol: TNFSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003807.4 | NP_003798.2 | 5 | 1077 | ||
NM_172014.3 | NP_742011.2 | 4 | 676 | |||
XM_017027417.1 | XP_016882906.1 | 4 | 1221 | |||
GenBank mRNA | AF036581.1 | - | 4 | 743 | ||
AF064090.1 | - | 5 | 1077 | |||
AK292037.1 | - | 4 | 786 | |||
AY028261.1 | - | 4 | 635 | |||
AY358812.1 | - | 4 | 753 | |||
BC018058.1 | - | 4 | 784 | |||
CR541854.1 | - | 4 | 695 | |||
CR541871.1 | - | 4 | 695 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
