siRNA Details
Gene Symbol: TNFSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003807.4 | NP_003798.2 | 2 | 479 | ||
NM_172014.3 | NP_742011.2 | 1 | 186 | |||
XM_017027417.1 | XP_016882906.1 | 1 | 623 | |||
XM_017027418.1 | XP_016882907.1 | 1 | 623 | |||
GenBank mRNA | AF036581.1 | - | 1 | 145 | ||
AF064090.1 | - | 2 | 479 | |||
AK292037.1 | - | 1 | 188 | |||
AY028261.1 | - | 1 | 145 | |||
AY358812.1 | - | 1 | 155 | |||
BC018058.1 | - | 1 | 186 | |||
CR541854.1 | - | 1 | 97 | |||
CR541871.1 | - | 1 | 97 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
