siRNA Details
Gene Symbol: CST11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080830.2 | NP_543020.2 | 1 | 114 | ||
NM_130794.1 | NP_570612.1 | 1 | 114 | |||
GenBank mRNA | AF335480.1 | - | 1 | 114 | ||
AF335481.1 | - | 1 | 114 | |||
BC121079.2 | - | 1 | 111 | 40119721 | ||
BC121080.2 | - | 1 | 111 | 40119722 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
