siRNA Details
Gene Symbol: HCST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014266.3 | NP_055081.1 | 3, 4 | 362 | ||
XM_017026193.1 | XP_016881682.1 | 3, 4 | 599 | |||
GenBank mRNA | AF122904.1 | - | 3, 4 | 304 | ||
AF172929.1 | - | 3, 4 | 339 | |||
AF285447.1 | - | 3, 4 | 279 | |||
AL050163.1 | - | 3, 4 | 362 | |||
BC035931.1 | - | 3, 4 | 313 | 4701500 | ||
BC046348.1 | - | 3, 4 | 362 | 5229680 | ||
BC065224.1 | - | 3, 4 | 318 | 5762412 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
