siRNA Details
Gene Symbol: KRT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002274.3 | NP_002265.2 | 7 | 1624 | ||
NM_153490.2 | NP_705694.2 | 8 | 1650 | |||
GenBank mRNA | AK092276.1 | - | 10 | 1579 | ||
AK223051.1 | - | 8 | 1649 | |||
AK223077.1 | - | 8 | 1649 | |||
BC002661.2 | - | 8 | 1634 | 3610361 | ||
BC077718.1 | - | 8 | 1627 | 5479174 | ||
X14640.1 | - | 8 | 1628 | |||
X52426.1 | - | 7 | 1608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
