siRNA Details
Gene Symbol: KRT13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002274.3 | NP_002265.2 | 1 | 163 | ||
NM_153490.2 | NP_705694.2 | 1 | 163 | |||
GenBank mRNA | AK092276.1 | - | 1 | 164 | ||
AK223051.1 | - | 1 | 162 | |||
AK223077.1 | - | 1 | 162 | |||
AK290244.1 | - | 1 | 162 | |||
AK309752.1 | - | 1 | 164 | |||
BC002661.2 | - | 1 | 147 | 3610361 | ||
BC077718.1 | - | 1 | 140 | 5479174 | ||
BC126184.1 | - | 1 | 127 | 8991900 | ||
BC136494.1 | - | 1 | 127 | 9020483 | ||
X14640.1 | - | 1 | 142 | |||
X52426.1 | - | 1 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
