siRNA Details
Gene Symbol: KRT35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002280.4 | NP_002271.3 | 8 | 1451 | ||
XM_011524794.1 | XP_011523096.1 | 7 | 3161 | |||
GenBank mRNA | X90763.1 | - | 8 | 1450 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
