siRNA Details
Gene Symbol: STATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003154.2 | NP_003145.1 | 3, 4 | 192 | ||
GenBank mRNA | AK311812.1 | - | 3, 4 | 157 | ||
BC067219.1 | - | 3, 4 | 158 | 30328926 | ||
BX649104.1 | - | 1 | 1900 | |||
CB958641.1 | - | 3, 4 | 159 | |||
CB986973.1 | - | 196 | ||||
M18078.1 | - | 3, 4 | 154 | |||
M18371.1 | - | 3, 4 | 138 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
