siRNA Details
Gene Symbol: STATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009181.1 | NP_001009181.1 | 4 | 233 | ||
NM_003154.2 | NP_003145.1 | 5 | 263 | |||
GenBank mRNA | AK311812.1 | - | 5 | 228 | ||
BC067219.1 | - | 5 | 229 | 30328926 | ||
BX649104.1 | - | 1 | 3024 | |||
CB958641.1 | - | 5 | 230 | |||
CB986973.1 | - | 267 | ||||
M18078.1 | - | 5 | 225 | |||
M18371.1 | - | 5 | 209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
