siRNA Details
Gene Symbol: STATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003154.2 | NP_003145.1 | 4, 5 | 215 | ||
GenBank mRNA | AK311812.1 | - | 4, 5 | 180 | ||
BC067219.1 | - | 4, 5 | 181 | 30328926 | ||
CB958641.1 | - | 4, 5 | 182 | |||
CB986973.1 | - | 219 | ||||
M18078.1 | - | 4, 5 | 177 | |||
M18371.1 | - | 4, 5 | 161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
