siRNA Details
Gene Symbol: HTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002159.3 | NP_002150.1 | 6 | 448 | ||
GenBank mRNA | BC017835.1 | - | 6 | 421 | 4289874 | |
BP335073.2 | - | 6 | 425 | |||
CB984538.1 | - | 6 | 452 | |||
CR627329.1 | - | 3 | 3605 | |||
M26664.1 | - | 5 | 363 |
Gene Symbol: SCYL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647352.1 | - | 2166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
