siRNA Details
Gene Symbol: HTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002159.3 | NP_002150.1 | 3, 4 | 176 | ||
GenBank mRNA | BC017835.1 | - | 3, 4 | 149 | 4289874 | |
BP335073.2 | - | 3, 4 | 153 | |||
CB984538.1 | - | 3, 4 | 180 | |||
CR627329.1 | - | 2 | 356 | |||
M26664.1 | - | 2, 3 | 91 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
