siRNA Details
Gene Symbol: GDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001492.5 | NP_001483.3 | 4 | 736 | ||
GenBank mRNA | M62302.1 | - | 4 | 688 |
Gene Symbol: CERS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290265.1 | NP_001277194.1 | 4 | 582 | ||
NM_021267.4 | NP_067090.1 | 4 | 736 | |||
NM_198207.2 | NP_937850.1 | 4 | 736 | |||
GenBank mRNA | AK293463.1 | - | 4 | 582 | ||
BC084582.1 | - | 4 | 534 | 6192817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
