siRNA Details
Gene Symbol: EVI2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003927.2 | NP_001003927.1 | 3 | 1180 | ||
NM_014210.3 | NP_055025.2 | 2 | 1039 | |||
GenBank mRNA | AK295311.1 | - | 3 | 1115 | ||
BC035572.1 | - | 2 | 927 | 5229926 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
