siRNA Details
Gene Symbol: PLA2G2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316722.1 | NP_001303651.1 | 2 | 124 | ||
XM_017001270.1 | XP_016856759.1 | 1 | 891 | |||
GenBank mRNA | AW135717.1 | - | 1 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
