siRNA Details
Gene Symbol: APOL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF305225.1 | - | 5 | 960 | ||
AF324223.1 | - | 5 | 1076 | |||
AF324224.1 | - | 6 | 1217 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
