siRNA Details
Gene Symbol: STRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134336.1 | NP_001127808.1 | 20 | 2116 | ||
NM_020704.2 | NP_065755.1 | 20 | 2116 | |||
XM_011516432.2 | XP_011514734.1 | 17 | 1764 | |||
XM_017012471.1 | XP_016867960.1 | 14 | 1439 | |||
XM_017012472.1 | XP_016867961.1 | 18 | 3581 | |||
GenBank mRNA | AB032996.1 | - | 20 | 2090 | ||
AK290165.1 | - | 20 | 2116 | |||
AK302339.1 | - | 12 | 1321 | |||
BC019064.2 | - | 20 | 2103 | 4590587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
