siRNA Details
Gene Symbol: STRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134336.1 | NP_001127808.1 | 7 | 659 | ||
NM_020704.2 | NP_065755.1 | 7 | 659 | |||
XM_006716069.3 | XP_006716132.1 | 7 | 659 | |||
XM_011516432.2 | XP_011514734.1 | 4 | 307 | |||
XM_011516435.1 | XP_011514737.1 | 7 | 659 | |||
XM_017012470.1 | XP_016867959.1 | 7 | 659 | |||
GenBank mRNA | AB032996.1 | - | 7 | 633 | ||
AK290165.1 | - | 7 | 659 | |||
BC019064.2 | - | 7 | 646 | 4590587 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
