siRNA Details
Gene Symbol: MTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006635.2 | NP_001006636.1 | 11 | 1187 | ||
NM_001319097.1 | NP_001306026.1 | 10 | 914 | |||
NM_001319098.1 | NP_001306027.1 | 9 | 906 | |||
NM_006554.4 | NP_006545.1 | 10 | 983 | |||
XM_011510497.2 | XP_011508799.1 | 11 | 1080 | |||
GenBank mRNA | AF053551.1 | - | 10 | 956 | ||
AK001287.1 | - | 10 | 935 | |||
AK289359.1 | - | 10 | 934 | |||
AK303564.1 | - | 11 | 1112 | |||
AK315918.1 | - | 11 | 1123 | |||
BC017271.2 | - | 11 | 1117 | 4635884 | ||
BC067831.1 | - | 10 | 962 | 5298149 | ||
BC088359.1 | - | 10 | 918 | 30378040 | ||
CX165048.1 | - | 10 | 807 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
