siRNA Details
Gene Symbol: MTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006635.2 | NP_001006636.1 | 11 | 1212 | ||
NM_001319097.1 | NP_001306026.1 | 10 | 939 | |||
NM_001319098.1 | NP_001306027.1 | 9 | 931 | |||
NM_006554.4 | NP_006545.1 | 10 | 1008 | |||
XM_011510497.2 | XP_011508799.1 | 11 | 1105 | |||
GenBank mRNA | AF053551.1 | - | 10 | 981 | ||
AK289359.1 | - | 10 | 959 | |||
AK303564.1 | - | 11 | 1137 | |||
AK315918.1 | - | 11 | 1148 | |||
BC017271.2 | - | 11 | 1142 | 4635884 | ||
BC067831.1 | - | 10 | 987 | 5298149 | ||
BC088359.1 | - | 10 | 943 | 30378040 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
