siRNA Details
Gene Symbol: WISP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204869.1 | NP_001191798.1 | 3 | 1119 | ||
NM_001204870.1 | NP_001191799.1 | 2 | 839 | |||
NM_003882.3 | NP_003873.1 | 5 | 1574 | |||
NM_080838.2 | NP_543028.1 | 4 | 1313 | |||
NR_037944.1 | 3 | 1033 | ||||
GenBank mRNA | AF100779.1 | - | 5 | 1556 | ||
AK301508.1 | - | 3 | 1033 | |||
KC344835.1 | - | 1 | 1588 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
